Learn More
Mouse TNF ELISPOT Pair, BD
Convenient set of unlabelled anti-TNF capture antibody and a biotinylated anti-TNF detection antibody
Brand:  BD Biosciences 551875
765.81 EUR valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 15846718
  • 770.00 EUR / Each
Estimated Shipment: 14-02-2025
to see stock.
Explore more special offers
Add to basket
Includes: Mouse TNF ELISPOT Capture Antibody (no azide/low endotoxin), Mouse TNF ELISPOT Detection Antibody (biotinylated) and a Certificate of Analysis that provides lot-specific optimal reagent concentrations