Learn More
MP Biomedicals™ Bovine Adult Serum
Used for the cultivation of Helicobacter pylori and in various low-serum media for diploid cell culture
Brand: MP Biomedicals™ 0219135280
Additional Details : Weight : 0.01000kg
Specifications
Golden Brown | |
Bovine | |
Adult Bovine Serum | |
Sterile | |
Cultivation of Helicobacter pylori and in various low Serum media for diploid cell culture | |
No | |
7.4 to 8.1 |
-20°C | |
Liquid | |
100 mL | |
0.2 μm filtration | |
12.4mg/dL | |
296mOsm | |
7.2 to 8.5g/dL |
Safety and Handling
- Chlorhexidine, free base
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P321-Specific treatment (see on this label).
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Chlorhexidine