missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ cappel™ LIPOPROTEIN

Filter sterilized, human high density lipoprotein (HDL)

Brand:  MP Biomedicals™ 0859433

Product Code. 11474465

  • 338.00 EUR / 10mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Isolated from fresh human serum by sequential isopycnic ultracentrifugation.

  • Density range for isolation is 1.063—1.21g/ml
  • Filtered through a 0.45μm filter and packaged aseptically
  • The identity and purity of each preparation is verified by cellulose acetate electrophoresis
  • The human serum used to derive the protein was negative when tested in accordance with current FDA required tests, and found to be non reactive for the presence of HIV 1and HIV 2 antibodies and hepatitis C virus (HCV), hepatitis B surface antigen (HBsAg), HIV 1 antigen and Syphilis

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Cell Biology and Analysis, Protein Analysis
7.4
Human Serum
Human
Human Low Density Lipoprotein
10mg
-40° to -70°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers