Learn More
MP Biomedicals™ Complement, Guinea Pig Purified, 5mL
Suitable for use in hemolytic titrations and cytotoxicity assays, and as a serum protein control in immunoelectrophoresis, Ouchterlony immunodiffusion, and other immunoprecipitation techniques
Brand:  MP Biomedicals™ 0855854
Additional Details : Weight : 0.00500kg
Product Code. 11444235
  • 150.52 EUR / 5mL
Estimated Shipment: 01-08-2024
Specifications
2° to 8°C
Guinea Pig complement
5 mL