Learn More
MP Biomedicals™ Dulbecco's MEM, 1X Liq., w/o L-Glut., L-Leucine
Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
Specifications
Specifications
| Form | Liquid |
| Product Type | DMEM Media |
| Sterility | Sterile |
| Without Additives | No Glutamine, No Leucine, No Sodium Pyruvate |
| Quantity | 100 mL |
| pH | 6.9 to 7.2 |
| Content And Storage | 2° to 8°C |