missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Goat Serum: Normal, Sterile
Suitable replacement for FBS on most cell lines and primary cultures for Biomedical Research
Brand: MP Biomedicals™ 0219135691
This item is not returnable.
View return policy
Description
Specifications
U.S. regulations | |
-20°C | |
Goat Serum | |
Sterile | |
Immunohistochemistry | |
No |
Red | |
Goat | |
1 L | |
0.2 μm filtration | |
<20mg/dL | |
6.5 to 8 |