missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Human Cytokines

Product Code. 11448001 Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 KU
Unit Size:
10ku
This item is not returnable. View return policy

Product Code. 11448001

Brand: MP Biomedicals™ 0215134210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Suitable for supplementing cell cultures and investigating various metabolic processes. Store as detailed on label.

TRUSTED_SUSTAINABILITY

Specifications

Reactivity Human, Mouse
Endotoxin Level 0.1ng/μg
Purity >97% by SDS-PAGE and HPLC analysis
Quantity 10 KU
Source E. coli
Sterility Sterile
Content And Storage -70°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.