missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Insulin Like Growth Factor 1, Human, Recombinant
This 70 amino acid peptide sequence is suitable for receptor binding assays, immunoassays, and cell culture applications.
Brand: MP Biomedicals™ 0215350525
This item is not returnable.
View return policy
Description
- Expressed in E. coli
- Endotoxin level: <0.1 ng/mg protein
Order Info
Supplied lyophilized
Specifications
-20°C | |
Lyophilized from a sterile-filtered solution in PBS | |
≥96% | |
E. coli |
Growth Factor | |
Receptor binding assays, immunoassays, and cell culture applications | |
25 μg | |
Human |