Learn More
MP Biomedicals™ Mouse Serum
Used for blocking agents and controls in immunoassays.
Products 3
Product Code Brand Form Hemoglobin Concentration Quantity Price Quantity & Availability  
Product Code Brand Form Hemoglobin Concentration Quantity Price Quantity & Availability  
11408541
View Documents
MP Biomedicals™
0215228201
Liquid 9.65 mg/dL 1 mL N/A
N/A
11418541
View Documents
MP Biomedicals™
0215228205
Liquid 9.65 mg/dL 5 mL N/A
N/A
11423976
View Documents
MP Biomedicals™
08642931
Lyophilized 5.8 g/dL 10 mL N/A
N/A