Learn More
Penicillin G Potassium Salt, MP Biomedicals™
Penicillin G Potassium Salt Cell culture reagent, White Powder
Brand: MP Biomedicals 0219453610
Additional Details : CAS Number : 113-98-4 Weight : 0.01000kg
Description
- It is used as a cell culture additive as an antibiotics.
- Penicillin G potassium Salt is a salt of Penicillin G.
- Penicillin G is a hydrophobic, β-lactam antibiotic produced by Penicillium spp.
- Penicillin G is also known as benzylpenicillin and contains a 6’ phenylacetyl side chain.
- In acid aqueous solutions, Penicillin is reported to decompose into penillic acid and penilloic acid.
Specifications
4°C | |
Antibiotic | |
10 μm |
Crystalline powder | |
Non-sterile |