missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Plant Culture Salt Mixtures

Plant Culture Salt Mixtures are for the molecular testing of plants and plant-derived samples for improving the productivity, quality, and disease resistance of plants.

Brand:  MP Biomedicals™ 092621622

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 11454895

  • 38.80 EUR / 10 L
Estimated Shipment: 05-08-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

  • Ready to use
  • Liquid formulation, prepared according to the original specification, except where noted
  • Available in different pack sizes, ready to use and liquid formulations
Specifications

Specifications

0°C to 5°C
Gamborg's Basal Salt Mixture
3.75 to 4.25
Powder
Solution
1 x 10 L
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers