Learn More
MP Biomedicals™ Pseudomonas Agar F
Used with glycerol (cat. no. 1004417) for the isolation, cultivation and differentiation of Pseudomonas aeruginosa on the basis of phycocyanin (pigment) production
Brand: MP Biomedicals™ 091008817
This item is not returnable.
View return policy
Description
Specifications
Room Temperature | |
Pseudomonas Agar F | |
Polipeptone 20g/L, Magnesium Sulfate 1.5g/L, Dipotassium Phosphate 1.5g/L, Agar 14g/L | |
500 g |
Dehydrated | |
Agar | |
Isolation, cultivation and differentiation of Pseudomonas aeruginosa |