missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Sephadex™ G-10
Fractiontionl Range (MW)
Brand: MP Biomedicals™ 0219525110
This item is not returnable.
View return policy
Description
- Globular proteins: <700
- Dextrans: <700
- Dry Bead Diameter: 40 to 120μm
- Bed Volume: 2 to 3mL/g
- CAS: 9050-68-4
Specifications
10 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction