Learn More
MP Biomedicals Sodium Deficient Diet, Rats and Mice
Based on AIN-76A
Brand:  MP Biomedicals 0296023210
 View more versions of this product
Product Code. 11407101
  • 583.00 EUR / 10kg
Description
This diet contains the normal chloride level.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Sodium Deficient Diet
Pellet
10,000 g
15°C to 30°C
Casein Low Trace Element 20%, DL-Methionine 0.3%, Corn Starch 15%, Sucrose 50%, Alphacel, Hydrolyzed 5%, Corn Oil 5%, Choline Bitartrate 0.2%, AIN-76 Mineral Mix (minus selenium) 3.5%, AIN-76a Vitamin Mix 1%
Powder
Rat