Learn More
MP Biomedicals Sodium Deficient Diet, Rats and Mice
Based on AIN-76A
Brand: MP Biomedicals 0296023210
This item is not returnable.
View return policy
Description
This diet contains the normal chloride level.
Specifications
Sodium Deficient Diet | |
Pellet | |
10,000 g | |
15°C to 30°C |
Casein Low Trace Element 20%, DL-Methionine 0.3%, Corn Starch 15%, Sucrose 50%, Alphacel, Hydrolyzed 5%, Corn Oil 5%, Choline Bitartrate 0.2%, AIN-76 Mineral Mix (minus selenium) 3.5%, AIN-76a Vitamin Mix 1% | |
Powder | |
Rat |