Learn More
MP Biomedicals™ SPIN Module and Recovery Tubes
Designed for optimal performance, providing the purest DNA faster than ever, and with no matrix carry-over. SPIN Module and Recovery Tubes are applied in molecular biology for their ability to separate particles from solutions in a few seconds.
Brand: MP Biomedicals™ 112080600
This item is not returnable.
View return policy
Description
- Spin Module give faster and cleaner separations than traditional methods such as columns, filters, or simple centrifugation
- When used with the GENECLEAN™ or MERmaid™ protocols, they combine the speed, efficiency, and selectivity of the GLASSMILK™ binding reaction with an even more rapid execution of the separation, washing, and elution steps
Specifications
Room Temperature | |
SPIN Module is applied in molecular biology for its ability to separate particles from solutions in a few seconds. | |
60 Pk. |
Lysate or other suspension | |
SPIN Module and Recovery Tubes | |
500 to 700 μL liquid per centrifugation, Larger volumes require multiple loadings |
Product Content Correction
Product Title
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction