missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Whole Serum, Chicken, Purified, 2mL

Whole Chicken Serum with buffer salts

Brand:  MP Biomedicals™ 0855982

Additional Details : Weight : 0.00200kg

Product Code. 17357459

  • 86.92 EUR / 2mL
Estimated Shipment: 16-08-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

2° to 8°C
Chicken Serum
0.2 μm filtration
Yes
61.9mg/mL
Chicken
2 mL
Serum Protein control in immunoelectrophoresis, Ouchterlony immunodiffusion, and other immunoprecipitation techniques, used as a blocking agent in fluorescence and enzyme immunoassays
7.3
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers