missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Whole Serum, Rabbit, Purified, 2mL

Whole Rabbit Serum with buffer salts

Brand:  MP Biomedicals™ 0855990

Product Code. 17561116

  • 89.57 EUR / 2mL
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

TRUSTED_SUSTAINABILITY
Specifications

Specifications

-20°C
Lyophilized
2 mL
Delipidated, Sodium Phosphate 0.02M, Sodium Chloride 0.14M, pH 7.3, Sodium Azide 0.05%
Yes
Rabbit
Rabbit Serum
62.9 mg/mL
Blocking assays, immunohistochemical
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers