missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Yeast Lytic Enzyme, from Arthrobacter luteus
Brand: MP Biomedicals™ 0215352650
This item is not returnable.
View return policy
Description
- Activity 20,000units/g
Specifications
20,000 U/g | |
7.5 | |
0°C | |
50 kU | |
Lyophilized Powder |
Tan | |
Used to digest the cell wall of active Yeast cells | |
Arthrobacter luteus | |
Yeast Lytic Enzyme |