Learn More
Welch™ MPC 301 Zp, ATEX Cat.2 Diaphragm Pump
Product Code. 10091832 Shop All Welch Ilmvac Products
Click to view available options
Flow Rate:
38 L/min.
Unit Size:
Each
Product Code. 10091832
Brand: Welch™ 400048104
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for use as a backing pump for rotary evaporators, vacuum ovens or gel dryer. Welch Ilmvac™ MPC 301 Zp Diaphragm Pumps are ideal for vacuum filtration, vacuum concentrating, distillation and desiccation.
  • ATEX Cat.2 explosion proof diaphragm pump
  • Manual Vacuum regulator with dial gauge and liquid trap
  • Powerful corrosion resistant diaphragm vacuum pump build-in
  • Maintenance-free drive system and proven long diaphragm life
  • Requires minimal benchtop space due to compact design
  • Analytically pure, oil free vacuum
  • Light weight, rugged design
  • All wetted parts are made of Ryton, Viton or Kalrez
  • Designed for continuous running
  • Use in the lab or in the field (with optional adapter)
  • CE Marked
TRUSTED_SUSTAINABILITY
Specifications
Electrical Requirements 400 V 50 Hz
Depth (Metric) 300 mm
Model MPC 301 Zp, ATEX Cat.2
Weight (Metric) 22.9 kg
Temperature (Metric) Max 40°C
Temperature (Metric) Min. 10°C
Flow Rate 38 L/min.
Frequency 50 Hz
Overall Height (Metric) 260 mm
Overall Width (Metric) 240 mm
Pump Type Diaphragm Pump
Maximum Pressure <8 mbar
Voltage 400 V