missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC3 Mouse anti-Human, Clone: rMUC3/1154, Novus Biologicals™
Mouse Monoclonal Antibody
255.00 EUR - 507.00 EUR
Specifications
Antigen | MUC3 |
---|---|
Clone | rMUC3/1154 |
Concentration | 0.2 mg/ml |
Dilution | Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Immunohistochemistry (Paraffin) |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18331674
|
Bio-Techne
NBP3-13973-20UG |
20 μg |
255.00 EUR
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18347454
|
Bio-Techne
NBP3-13973-100UG |
100 μg |
507.00 EUR
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MUC3 Monoclonal antibody specifically detects MUC3 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
MUC3 | |
0.2 mg/ml | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Human | |
Q02505, Q9H195 | |
4584 | |
IgG2a κ | |
Protein A or G purified |
rMUC3/1154 | |
Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
10 mM PBS with 0.05% BSA | |
MUC-3A, mucin 3, intestinal, mucin 3A, cell surface associated, mucin 3A, intestinal, mucin-3A | |
Recombinant fragment of human MUC3A protein (exact sequence is proprietary) | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title