Learn More
Velp Scientifica™ MultiAluBlock™ Base
Shop All Velp Products
Click to view available options
For Use With (Equipment):
Series 6 Hotplate Stirrers
Unit Size:
Each
Description
- Used to perform numerous reactions reducing time and bench space
- For use with Velp Series 6 Hotplate Stirrers
Specifications
Specifications
| Description | Holds various combinations of MultiAluBlocks™ |
| Diameter (Metric) | 135 mm |
| For Use With (Equipment) | Series 6 Hotplate Stirrers |
| Type | Base |