Learn More
Fisherbrand™ Aluminium Adapter with Wrench
Provides a stable base able to be extended at right angles to fit scaffolding feet. Fisherbrand™ Scaffolding Foot Adapters accept 3 x 13mm diameter rods or tubes at right angles - two perpendicular to the boss, the third in line with the boss.
Brand:  Fisherbrand™ 2119
Product Code. 11598112
  • 10.80 EUR / Each
Estimated Shipment: 31-03-2025
Description
  • With Allen key and boss
  • With socket head screw fixings for rods or tubes
  • Manufactured from durable stainless steel
Note: For use with scaffolding feet (11578112).
Specifications
Specifications
Multiple Connector
For use with scaffolding feet 11578112.