Learn More
Axygen™ MultiRack Empty Rack
Product Code. p-7159933 Shop All Axygen Scientific Products
Click to view available options
For Use With (Equipment):
10 μL XT, 200 μL, 200 μL NX, 300 μL
200 μL XT, 200 μL XT, 300 μL XT, 1000 μL XT
Unit Size:
Pack of 5
Product Code. 16372237
Brand: Axygen™ MRRA
Only null left
Add to basket
Add to basket
MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in Bulk, Racked, Filter and Reload packaging styles to meet the variety of needs in the lab setting.
  • Detectable DNase/RNase/Human DNA/PCR inhibitor free
  • Nonpyrogenic
  • Manufactured in an ISO certified facility in the USA
  • Fully autoclavable
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ISO 13845
Autoclavable Autoclavable
Color Clear
Material Polyethylene
Sterility Non-sterile
For Use With (Equipment) 10 μL XT, 200 μL, 200 μL NX, 300 μL
No. per Case 1/Cs.
No. per Pack 5/Pk.
Quantity 5/pk, 1 pk/cs