Learn More
Lonza™ Mycoalert™ Mycoplasma Detection Kit
Detect mycoplasma in culture. Mycoalert™ Mycoplasma Detection Kit is a selective biochemical test that exploits the activity of mycoplasmal enzymes which are found in all six of the main mycoplasma cell culture contaminants and the vast majority of 180 mycoplasma species, but are not present in eukaryotic cells.
Brand: Lonza™ LT07-318
This item is not returnable.
View return policy
Description
Convenient & fast- Simply add two reagents to the culture supernatant
- Perform two luminescence readings
- Results in < 20 min
- Easy interpretation of results
- Detects all common mollicute contaminations (Mycoplasma, Acholeplasma, Entomoplasma and Spiroplasma; except Ureaplasma)
- Negligible interference with media components
Specifications
Mycoalert reagent: 1 x 10mL, Mycoalert assay buffer: 1 x 20mL, Mycoalert substrate: 1 x 10mL 2 to 8°C |
|
Mammalian Cells | |
Mycoplasma Detection | |
100 Tests |
Safety and Handling
Hazard Statement
- EUH210-Safety data sheet available on request.