Learn More
Thermo Scientific™ mySPIN™ 6 Mini Centrifuge
Product Code. p-9367001 Shop All Thermo Scientific Products
Click to view available options
Type:
12 V Adapter
Benchtop Mini Centrifuge
Transport Box with Inserts
Unit Size:
Each
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Product Code. 15334204
Brand: Thermo Scientific™ 75004061
Availability and estimated dispatch date are for standard equipment. If factory fitted options, customisation or services are required, please Request A Quote or contact your Account Manager as lead times will vary.
Our most compact centrifuge, the mySPIN 6 mini centrifuge, is ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microcentrifuge tubes.
Bring efficiency and user comfort to your lab stations with the Thermo Scientific mySPIN 6 mini centrifuge. Ergonomic design, tool-free rotors, and quick-spin options make it fast and easy to use. Ideal for microfilter cell separations and HPLC samples, with adapters to accommodate all major microtube and PCR tube styles (2.0, 1.5, 0.5, 0.2 mL) for customized use.
Enjoy convenience at your personal workstation with our most compact centrifuges, featuring:
  • Small footprint for efficient use of lab space
  • Easy operation and palm-shaped lid for left- or right-handed users
  • Quick-spin feature
  • Quiet operation, low vibration
  • Safety features including unit will not operate without lid in place for safety
  • Includes 2 rotors to accommodate 0.2, 0.5, 1.5, 2.0 mL microcentrifuge tubes and PCR strip tubes
  • Tool-free, quick rotor exchange
TRUSTED_SUSTAINABILITY
Specifications
Capacity 6 x 2 mL
Max. Speed 6,000 rpm
Max. RCF 2,000 x g
Noise Level 51-53 dB(A)
Temperature Range Ambient temperature
Certifications/Compliance CSA, CE, UL
Type Benchtop Mini Centrifuge
Includes 6 x 1.5/2.0mL tube rotor, 16 x 0.2mL tube rotor (singles or 2 x 8 strip), 6 tube adapters 0.2mL, 6 tube adapters 0.5mL, storage case for rotors and adapters, tube rack, universal power adapter 110-240V 50/60 Hz, with 4 interchangeable plug type
Electrical Requirements 110/240 V 50/60 Hz with 4 interchangeable plug adapters
Warranty 2 Years
Refrigerated No
Standards IEC 61010-1, IEC 61010-2-020 (refer to manual for more details)
Dimensions (HWD) 4.1 x 5.0 x 6.0 in. (10.4 x 12.8 x 15.3 cm)
Net Weight 1.6 lb. (0.74 kg)
Product Line mySpin 6
Frequency 50/60 Hz
Voltage 110/240 V
Show More Show Less
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado