missing translation for 'onlineSavingsMsg'
Learn More

N-(1-Benzothien-2-ylmethyl)-N-methylamine hydrochloride, ≥97%, Thermo Scientific™

90.20 EUR - 402.00 EUR

Chemical Identifiers

CAS 849776-43-8
Molecular Formula C10H12ClNS
Molecular Weight (g/mol) 213.72
MDL Number MFCD03306023
InChI Key PBEZSHRKTUDPLK-UHFFFAOYSA-N
Synonym n-1-benzothien-2-ylmethyl-n-methylamine hydrochloride, 1-benzo b thiophen-2-yl-n-methylmethanamine hydrochloride, 1-benzothien-2-ylmethyl methylamine hydrochloride, 1-benzothiophen-2-ylmethyl methyl amine hydrochloride, benzo b thiophen-2-ylmethyl-methyl-ammonium chloride, c10h11ns.hcl, benzo b thiophen-2-ylmethyl methylamine, chloride, 1-benzo b thiophen-2-yl ;-n-methylmethanamine hydrochloride, 1-1-benzothiophen-2-yl-n-methylmethanamine-hydrogen chloride 1/1
PubChem CID 16956076
SMILES Cl.CNCC1=CC2=CC=CC=C2S1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10780334
View Documents Promotion Details
Thermo Scientific Maybridge
CC12296CB
250 mg
90.20 EUR
250mg
Please to purchase this item. Need a web account? Register with us today!
10147842
View Documents Promotion Details
Thermo Scientific Maybridge
CC12296DA
1 g
124.00 EUR
1g
Please to purchase this item. Need a web account? Register with us today!
10163164
View Documents Promotion Details
Thermo Scientific Maybridge
CC12296DE
5 g
402.00 EUR
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

849776-43-8
213.72
PBEZSHRKTUDPLK-UHFFFAOYSA-N
16956076
C10H12ClNS
MFCD03306023
n-1-benzothien-2-ylmethyl-n-methylamine hydrochloride, 1-benzo b thiophen-2-yl-n-methylmethanamine hydrochloride, 1-benzothien-2-ylmethyl methylamine hydrochloride, 1-benzothiophen-2-ylmethyl methyl amine hydrochloride, benzo b thiophen-2-ylmethyl-methyl-ammonium chloride, c10h11ns.hcl, benzo b thiophen-2-ylmethyl methylamine, chloride, 1-benzo b thiophen-2-yl ;-n-methylmethanamine hydrochloride, 1-1-benzothiophen-2-yl-n-methylmethanamine-hydrogen chloride 1/1
Cl.CNCC1=CC2=CC=CC=C2S1

Specifications

849776-43-8
100.0
Amber Glass Bottle
MFCD03306023
PBEZSHRKTUDPLK-UHFFFAOYSA-N
[(1-benzothiophen-2-yl)methyl](methyl)amine hydrochloride
16956076
97%
96.5
97%
C10H12ClNS
n-1-benzothien-2-ylmethyl-n-methylamine hydrochloride, 1-benzo b thiophen-2-yl-n-methylmethanamine hydrochloride, 1-benzothien-2-ylmethyl methylamine hydrochloride, 1-benzothiophen-2-ylmethyl methyl amine hydrochloride, benzo b thiophen-2-ylmethyl-methyl-ammonium chloride, c10h11ns.hcl, benzo b thiophen-2-ylmethyl methylamine, chloride, 1-benzo b thiophen-2-yl ;-n-methylmethanamine hydrochloride, 1-1-benzothiophen-2-yl-n-methylmethanamine-hydrogen chloride 1/1
Cl.CNCC1=CC2=CC=CC=C2S1
213.72
213.73
N-(1-Benzothien-2-ylmethyl)-N-methylamine hydrochloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

SDS
Documents

Documents

Product Certifications