missing translation for 'onlineSavingsMsg'
Learn More

N-(2-Ethoxyphenyl)-2-methoxybenzamide, 97%, Thermo Scientific™

Product Code. 15476208
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 306279-65-2
Molecular Formula C16H17NO3
Molecular Weight (g/mol) 271.316
MDL Number MFCD01010708
InChI Key IFOHCGGZEGVBME-UHFFFAOYSA-N
Synonym n-2-ethoxyphenyl-2-methoxybenzamide, n-2-ethoxyphenyl 2-methoxyphenyl carboxamide
PubChem CID 700574
IUPAC Name N-(2-ethoxyphenyl)-2-methoxybenzamide
SMILES CCOC1=CC=CC=C1NC(=O)C2=CC=CC=C2OC

Specifications

Quantity 250 mg
Formula Weight 271.32
Percent Purity 97%
Chemical Name or Material N-(2-Ethoxyphenyl)-2-methoxybenzamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.