missing translation for 'onlineSavingsMsg'
Learn More

N-[3-(Trifluoromethyl)phenylsulfonyl]-DL-phenylalanine, 96%, Thermo Scientific™

Product Code. 15428525
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15428525

Brand: Thermo Scientific Alfa Aesar H33543.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 250714-63-7
Molecular Formula C16H14F3NO4S
Molecular Weight (g/mol) 373.35
MDL Number MFCD00173005
InChI Key JFDJLMGVJXHLOF-UHFFFAOYNA-N
Synonym 3-phenyl-2-3-trifluoromethyl phenyl sulfonylamino propanoic acid, 3-phenyl-2-3-trifluoromethyl benzenesulfonamido propanoic acid, 3-phenyl-2-3-trifluoromethyl phenyl sulfonyl amino propanoic acid, 3-trifluoromethyl phenyl sulfonyl phenylalanine, phenylalanine,n-3-trifluoromethyl phenyl sulfonyl, n-3-trifluoromethyl phenyl sulphonyl-dl-phenylalanine, 3-phenyl-2-3-trifluoromethyl phenylsulfonamido propanoic acid, 3-phenyl-2-3-trifluoromethyl phenyl-sulfonyl amino propanoic acid
PubChem CID 2776443
IUPAC Name 3-phenyl-2-[[3-(trifluoromethyl)phenyl]sulfonylamino]propanoic acid
SMILES OC(=O)C(CC1=CC=CC=C1)NS(=O)(=O)C1=CC=CC(=C1)C(F)(F)F

Specifications

Melting Point 130°C to 135°C
Quantity 250 mg
Formula Weight 373.35
Percent Purity 96%
Chemical Name or Material N-[3-(Trifluoromethyl)phenylsulfonyl]-DL-phenylalanine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.