missing translation for 'onlineSavingsMsg'
Learn More

N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester, 99%, Thermo Scientific™

Product Code. 11485986
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11485986

Brand: Thermo Scientific Alfa Aesar B22305.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13726-52-8
Molecular Formula C16H22N2O5
Molecular Weight (g/mol) 322.361
MDL Number MFCD00009617
InChI Key RJXFBLRRPYBPTM-ZDUSSCGKSA-N
Synonym n-4-aminobenzoyl-l-glutamic acid diethyl ester, diethyl n-4-aminobenzoyl-l-glutamate, diethyl n-p-aminobenzoyl-l-glutamate, 1,5-diethyl 2s-2-4-aminophenyl formamido pentanedioate, 4-abz-glu oet-oet, l-glutamic acid, n-4-aminobenzoyl-, diethyl ester, diethyl 4-aminobenzoyl-l-glutamate, diethyl p-aminobenzoyl-l-glutamate, s-diethyl 2-4-aminobenzamido pentanedioate, diethyl 2-4-aminobenzoyl amino pentanedioate #
PubChem CID 73002
IUPAC Name diethyl (2S)-2-[(4-aminobenzoyl)amino]pentanedioate
SMILES CCOC(=O)CCC(C(=O)OCC)NC(=O)C1=CC=C(C=C1)N

Specifications

Melting Point 139°C to 142°C
Quantity 50 g
Formula Weight 322.36
Optical Rotation 19° (c=1 Chloroform)
Percent Purity 99%
Chemical Name or Material N-(4-Aminobenzoyl)-L-glutamic acid diethyl ester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.