missing translation for 'onlineSavingsMsg'
Learn More

N-acetyl-D-galactosamine, MP Biomedicals™

Product Code. 11368349
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

N-Acetyl-D-galactosamine(GalNAc), an aminosugar, is a component of many O-linked and N-linked glycan structures

TRUSTED_SUSTAINABILITY

Specifications

CAS 14215-68-0
Color White
Molecular Formula C8H15NO6
MDL Number MFCD00065372
Quantity 25 g
Synonym n-acetyl-d-galactosamine, galnac, 2-acetamido-2-deoxy-d-galactopyranose, d-galnac, 2-acetamido-2-deoxy-d-galactose, n-acetyl-d-chondrosamine, n-3r,4r,5r,6r-2,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-3-yl acetamide, n-acetylgalactosamine, acetylgalactosamine, n-acetyl-galactosamine
InChI Key OVRNDRQMDRJTHS-JAJWTYFOSA-N
SMILES CC(=O)NC1C(C(C(OC1O)CO)O)O
IUPAC Name N-[(2R,3R,4R,5R,6R)-2,4,5-trihydroxy-6-(hydroxymethyl)oxan-3-yl]acetamide
Molecular Weight (g/mol) 221.209
PubChem CID 440552
ChEBI CHEBI:28497
Formula Weight 221.21
Physical Form Powder
Chemical Name or Material N-acetyl-D-galactosamine
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'