missing translation for 'onlineSavingsMsg'
Learn More

N-Acetyl-D-Glucosamine, Cell Cculture Reagent, MP Biomedicals™

Product Code. 11425022
Click to view available options
Quantity:
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Specifications

CAS 72-87-7
Molecular Formula C8H15NO6
Quantity 100 g
Synonym n-acetyl-d-glucosamine, glcnac, 2-acetamido-2-deoxy-d-glucopyranose, 2-acetamido-2-deoxy-d-glucose, n-acetylchitosamine, n-acetylglucosamine, aflexa, d-glcnac, glucosamine complex, maxi gs
InChI Key OVRNDRQMDRJTHS-YQOGUDIHNA-N
Molecular Weight (g/mol) 221.21
PubChem CID 439174
ChEBI CHEBI:506227
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.