Learn More
N-Benzylacrylamide, 96%, Thermo Scientific Chemicals
Product Code. 11327776
Click to view available options
Quantity:
1 g
5 g
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 13304-62-6
Molecular Formula C10H11NO
Molecular Weight (g/mol) 161.204
MDL Number MFCD00015333
InChI Key OHLHOLGYGRKZMU-UHFFFAOYSA-N
Synonym n-benzylacrylamide, 2-propenamide, n-phenylmethyl, n-benzyl acrylamide, acmc-20ak6s, enamine_005841
PubChem CID 139428
IUPAC Name N-benzylprop-2-enamide
SMILES C=CC(=O)NCC1=CC=CC=C1
Specifications
Melting Point 59°C to 62°C
Quantity 1 g
Beilstein 2690053
Formula Weight 161.21
Percent Purity 96%
Chemical Name or Material N-Benzylacrylamide