missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyloxycarbonyl-L-aspartic acid 4-benzyl ester, 98+%, Thermo Scientific™

Product Code. 11368165
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is widely used as a pharmaceutical intermediate and in food industry.

Solubility
Insoluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3479-47-8
Molecular Formula C19H19NO6
Molecular Weight (g/mol) 357.36
MDL Number MFCD00037820
InChI Key VUKCNAATVIWRTF-UHFFFAOYNA-N
Synonym z-asp obzl-oh, cbz-asp obzl-oh, n-cbz-l-aspartic acid 4-benzyl ester, z-l-aspartic acid 4-benzyl ester, n-benzyloxycarbonyl-l-aspartic acid 4-benzyl ester, 4-benzyl hydrogen n-phenylmethoxy carbonyl-l-aspartate, s-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid, n-carbobenzoxy-l-aspartic acid 4-benzyl ester, 2s-4-oxo-4-phenylmethoxy-2-phenylmethoxycarbonylamino butanoic acid, 2s-4-benzyloxy-2-benzyloxy carbonyl amino-4-oxobutanoic acid
PubChem CID 47602
IUPAC Name (2S)-4-oxo-4-phenylmethoxy-2-(phenylmethoxycarbonylamino)butanoic acid
SMILES OC(=O)C(CC(=O)OCC1=CC=CC=C1)NC(=O)OCC1=CC=CC=C1

Specifications

Melting Point 105°C to 107°C
Quantity 1 g
Beilstein 2065292
Solubility Information Insoluble in water.
Formula Weight 357.37
Percent Purity ≥98%
Chemical Name or Material N-Benzyloxycarbonyl-L-aspartic acid 4-benzyl ester
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'