missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyltrifluoromethanesulfonamide, 97%, Thermo Scientific Chemicals

Product Code. 11399335
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36457-58-6
Molecular Formula C8H8F3NO2S
Molecular Weight (g/mol) 239.21
MDL Number MFCD00042074
InChI Key IJHVVEQTOXFGCL-UHFFFAOYSA-N
PubChem CID 118985
IUPAC Name N-benzyl-1,1,1-trifluoromethanesulfonamide
SMILES FC(F)(F)S(=O)(=O)NCC1=CC=CC=C1

Specifications

Melting Point 44°C to 46°C
Linear Formula C6H5CH2NHSO2CF3
Quantity 5 g
Beilstein 2119124
Synonym n-benzyltrifluoromethanesulfonamide, n-benzyltriflamide, n-benzyl trifluoromethanesulfonamide, n-benzyl-1,1,1-trifluoromethanesulphonamide, benzyl trifluoromethyl sulfonyl amine, trifluoromethanesulfonamidenbenzyl, acmc-1ahjf, 1,1,1-trifluoro-n-phenylmethyl methanesulfonamide, methanesulfonamide,1,1,1-trifluoro-n-phenylmethyl, methanesulfonamide, 1,1,1-trifluoro-n-phenylmethyl
Formula Weight 239.22
Percent Purity 97%
Chemical Name or Material N-Benzyltrifluoromethanesulfonamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'