missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-methyl-L-serine, 97%

Product Code. 15409607
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 15409607

Brand: Thermo Scientific Alfa Aesar H52829.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 51293-47-1 | C9H17NO5 | 219.237 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51293-47-1
Molecular Formula C9H17NO5
Molecular Weight (g/mol) 219.237
MDL Number MFCD00153314
InChI Key RFGMSGRWQUMJIR-LURJTMIESA-N
Synonym boc-o-methyl-l-serine, boc-ser me-oh, s-2-tert-butoxycarbonyl amino-3-methoxypropanoic acid, boc-ser me-oh.dcha, n-boc-o-methyl-l-serine, s-2-tert-butoxycarbonylamino-3-methoxypropanoic acid, boc-ser me-oh dcha, boc-o-methyl-l-ser, serine, n-1,1-dimethylethoxy carbonyl-o-methyl, 2s-2-tert-butoxycarbonyl amino-3-methoxypropanoic acid
PubChem CID 7016354
IUPAC Name (2S)-3-methoxy-2-[(2-methylpropan-2-yl)oxycarbonylamino]propanoic acid
SMILES CC(C)(C)OC(=O)NC(COC)C(=O)O

Specifications

Quantity 1 g
Formula Weight 219.24
Percent Purity 97%
Chemical Name or Material N-Boc-O-methyl-L-serine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.