Learn More
n-Butylethylmagnesium, 0.9M solution in heptane, AcroSeal™, Thermo Scientific Chemicals
122.00 EUR - 758.00 EUR
Chemical Identifiers
CAS | 62202-86-2, 142-82-5 |
---|---|
Molecular Formula | C6H14Mg |
Molecular Weight (g/mol) | 110.48 |
MDL Number | MFCD00072569 |
InChI Key | MVECFARLYQAUNR-UHFFFAOYSA-N |
Synonym | magnesium, butylethyl, butylethylmagnesium, n-butylethylmagnesium, butyl ethyl magnesium, n-butyl ethyl magnesium, mvecfarlyqaunr-uhfffaoysa-n |
PubChem CID | 112818 |
IUPAC Name | magnesium;butane;ethane |
SMILES | CCCC[Mg]CC |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10557303
|
Thermo Scientific Acros
381181000 |
100 mL | AcroSeal™ Glass Bottle |
122.00 EUR
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10778833
|
Thermo Scientific Acros
381188000 |
800 mL | AcroSeal™ Glass Bottle |
758.00 EUR
800mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
62202-86-2, 142-82-5 | |
110.48 | |
MVECFARLYQAUNR-UHFFFAOYSA-N | |
112818 | |
CCCC[Mg]CC |
C6H14Mg | |
MFCD00072569 | |
magnesium, butylethyl, butylethylmagnesium, n-butylethylmagnesium, butyl ethyl magnesium, n-butyl ethyl magnesium, mvecfarlyqaunr-uhfffaoysa-n | |
magnesium;butane;ethane |
Specifications
62202-86-2, 142-82-5 | |
0.7300g/mL | |
CH3CH2Mg(CH2)3CH3 | |
0.73 | |
Solubility in water: reacts | |
CCCC[Mg]CC | |
110.48 | |
110.48 | |
n-Butylethylmagnesium, 0.9M Solution in Heptane |
Colorless | |
C6H14Mg | |
MFCD00072569 | |
magnesium, butylethyl, butylethylmagnesium, n-butylethylmagnesium, butyl ethyl magnesium, n-butyl ethyl magnesium, mvecfarlyqaunr-uhfffaoysa-n | |
MVECFARLYQAUNR-UHFFFAOYSA-N | |
magnesium;butane;ethane | |
112818 | |
Liquid |
Safety and Handling
GHS H Statement
May cause drowsiness or dizziness.
Causes severe skin burns and eye damage.
May be fatal if swallowed and enters airways.
Very toxic to aquatic life with long lasting effects.
In contact with water releases
GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 263-454-
RUO – Research Use Only