missing translation for 'onlineSavingsMsg'
Learn More

N-(Diphenylmethylene)glycine tert-butyl ester, 99%

Product Code. 15415467
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15415467

Brand: Thermo Scientific Alfa Aesar H32709.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 81477-94-3 | C19H21NO2 | 295.382 g/mol

N-(Diphenylmethylene)glycine tert-butyl ester is a useful dye for biological research purposes. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-(Diphenylmethylene)glycine tert-butyl ester is a useful dye for biological research purposes. It is an important raw material and intermediate used in organic synthesis, pharmaceuticals agrochemicals and dyestuff fields.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81477-94-3
Molecular Formula C19H21NO2
Molecular Weight (g/mol) 295.382
MDL Number MFCD00134280
InChI Key YSHDPXQDVKNPKA-UHFFFAOYSA-N
Synonym n-diphenylmethylene glycine tert-butyl ester, diphenylmethylene-glycine t-butyl ester, tert-butyl 2-diphenylmethylene amino acetate, n-diphenylmethylene glycerine tert-butyl ester, tert-butyl 2-diphenylmethyleneamino acetate, diphenylmethyleneamino acetic acid tert-butyl ester, tert-butyl diphenylmethyleneamino acetate, n-diphenylmethylene glycinetert-butylester, tert-butyl 2-benzhydrylideneamino acetate, glycine, n-diphenylmethylene-, 1,1-dimethylethyl ester
PubChem CID 688171
IUPAC Name tert-butyl 2-(benzhydrylideneamino)acetate
SMILES CC(C)(C)OC(=O)CN=C(C1=CC=CC=C1)C2=CC=CC=C2

Specifications

Melting Point 113°C to 115°C
Linear Formula (C6H5)2C=NCH2COOC(CH3)3
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 295.38
Percent Purity 99%
Chemical Name or Material N-(Diphenylmethylene)glycine tert-butyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.