Learn More
N-Formyl-L-leucine, tech. 90%
Product Code. 15406397
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15406397
Brand: Thermo Scientific Alfa Aesar H33564.06
Request Bulk or Custom Format
CAS: 6113-61-7 | C7H13NO3 | 159.19 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6113-61-7
Molecular Formula C7H13NO3
Molecular Weight (g/mol) 159.19
MDL Number MFCD00055861
InChI Key HFBHOAHFRNLZGN-MDOHGIEYNA-N
Synonym n-formyl-l-leucine, n-formyl-leu-oh, n-formylleucine, l-leucine, n-formyl, unii-5tha63y4ff, 5tha63y4ff, 2s-2-formamido-4-methylpentanoic acid, n-formylleucine #, formylleucin, n-formyl leucine
PubChem CID 9880216
IUPAC Name (2S)-2-formamido-4-methylpentanoic acid
SMILES CC(C)C[C@H](NC=O)C(O)=O
Specifications
Melting Point 142°C to 145°C
Quantity 5 g
Formula Weight 159.18
Percent Purity 90%
Grade Technical
Chemical Name or Material N-Formyl-L-leucine
compliance-icons
Product Identifier
  • N-Formyl-L-leucine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : N-Formyl-L-leucine