missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Heptyl 4-hydroxybenzoate, 98%
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
n-Heptyl 4-hydroxybenzoate is used for inhibiting microbiological spoilage, used as preservatives, skin care, hair care.
Solubility
Soluble in alcohol, phenoxyethanol, propylene glycol. Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
n-Heptyl 4-hydroxybenzoate is used for inhibiting microbiological spoilage, used as preservatives, skin care, hair care.
Solubility
Soluble in alcohol, phenoxyethanol, propylene glycol. Insoluble in water.
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Melting Point | 46°C to 49°C |
| Quantity | 100 g |
| Solubility Information | Soluble in alcohol,phenoxyethanol,propylene glycol. Insoluble in water. |
| Formula Weight | 236.31 |
| Percent Purity | 98% |
| Chemical Name or Material | n-Heptyl 4-hydroxybenzoate |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction