missing translation for 'onlineSavingsMsg'
Learn More

n-Hexane, MP Biomedicals™

Product Code. 11461142 Shop All MP Biomedicals Products
Click to view available options
Quantity:
0.5 mL
100 mL
250 mL
1 L
Unit Size:
1 L
100mL
250mL
500mL
This item is not returnable. View return policy

Product Code. 11461142

Brand: MP Biomedicals 0219522080

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

n-Hexane, ≥99%, Liquid

  • Hexadecane has been used as a phase change material (PCM) in the preparation of hexadecane/xGnP (exfoliated graphite nanoplatelet), a composite PCM loaded with xGnP. Hexadecane (n-Hexadecane) is an aliphatic long-chain hydrocarbon. It is a reference fuel with the cetane number 100 and is employed in determining the cetane number of diesel fuels. The mechanism of the oxidation of hexadecane in the gas-phase has been described. The density, speed of sound, adiabatic compressibility and compressed liquid density of hexadecane have been determined. The performance of platinum/zeolite catalysts for the hydroisomerization of n-hexadecane has been assessed. The thermal degradation of n-hexadecane forms n-alkanes and 1-alkenes as key products.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-54-3
Molecular Formula C6H14
Molecular Weight (g/mol) 86.18
MDL Number MFCD02179311
InChI Key VLKZOEOYAKHREP-UHFFFAOYSA-N
PubChem CID 8058
ChEBI CHEBI:29021
IUPAC Name hexane
SMILES CCCCCC

Specifications

Melting Point -95°C (Literature)
Color 1 (APHA)
Density 0.66 at 20°C (water = 1); 0.659g/mL at 25°C(Lit.)
Boiling Point 69°C (Literature)
Quantity 100 mL
Physical Form Liquid
Refractive Index n20/D 1.375(Lit.)
Assay Percent Range 99.9%
Residue after Evaporation <0 ppm
Specific Gravity 0.674
Vapor Pressure 124 mm Hg at 20°C 150 mm Hg at 24.8°C; 256 mmHg ( 37.7°C), 5.2 psi ( 37.7°C), ∽132 mmHg ( 20°C)(Lit.)
UN Number UN1208
Beilstein 1730733
Synonym n-hexane, esani, skellysolve b, gettysolve-b, hexyl hydride, hexan, dipropyl, heksan, hexanen, hexanes
Solubility Information Soluble in water.
Formula Weight 86.18
Chemical Name or Material n-Hexane, For ACS analysis
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.