missing translation for 'onlineSavingsMsg'
Learn More
N-Methyl-N-[(5-morpholin-4-ylthien-2-yl)methyl]amine, 97%, Thermo Scientific™ Product Code.: 11356785

N-Methyl-N-[(5-morpholin-4-ylthien-2-yl)methyl]amine, 97%, Thermo Scientific™

Product Code. 11356785
250 mg, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 11356785

Brand: Thermo Scientific Maybridge CC53446CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 879896-62-5
Molecular Formula C10H16N2OS
Molecular Weight (g/mol) 212.311
MDL Number MFCD09025876
InChI Key LTBKEKXCWIEXPM-UHFFFAOYSA-N
Synonym n-methyl-n-5-morpholin-4-ylthien-2-yl methyl amine, methyl 5-morpholin-4-yl thiophen-2-yl methyl amine, methyl 5-morpholin-4-yl 2-thienyl methyl amine, 2-thiophenemethanamine,n-methyl-5-4-morpholinyl, n-methyl-1-5-morpholin-4-yl thiophen-2-yl methanamine
PubChem CID 18525863
IUPAC Name N-methyl-1-(5-morpholin-4-ylthiophen-2-yl)methanamine
SMILES CNCC1=CC=C(S1)N2CCOCC2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 212.32
Percent Purity 97%
Chemical Name or Material N-methyl-N-[(5-morpholin-4-ylthien-2-yl)methyl]amine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
N-Methyl-N-[(5-morpholin-4-ylthien-2-yl)methyl]amine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.