missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N,N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid, free acid, MP Biomedicals™
Click to view available options
Quantity:
100 g
250 g
1 kg
Specifications
Specifications
Color | White |
Physical Form | Crystalline Powder |
Quantity | 100 g |
Assay Percent Range | 0.9961 |
Formula Weight | 239.23 |
Chemical Name or Material | N, N-bis(2-Hydroxyethyl)-2-aminoethanesulfonic acid, free acid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction