Learn More
Melford n-Octylglucoside
Product Code. 30018550
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
Unit Size:
10g
1g
25g
50g
5g
Product Code. 30018550
Brand: Melford N0200725.0
Non-ionic detergent for the solubilization and isolation of membrane bound proteins.
Non-ionic detergent for the solubilization and isolation of membrane bound proteins.
Specifications
CAS 29836-26-8
Molecular Formula C14H28O6
Quantity 25 g