Learn More
Melford n-Octylglucoside
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
50 g
Unit Size:
10g
1g
25g
50g
5g
Description
Non-ionic detergent for the solubilization and isolation of membrane bound proteins.
Non-ionic detergent for the solubilization and isolation of membrane bound proteins.
Specifications
Specifications
| CAS | 29836-26-8 |
| Molecular Formula | C14H28O6 |
| Quantity | 5 g |