Learn More
3M™ Nailsaver Sponges
Product Code. 17118660
Click to view available options
Color:
Blue, Orange
Blue, White
Green, White
Pink
Red, White
Yellow, Green
Yellow, White
Dimensions (L x W):
92 x 70 mm
130 x 70 mm
150 x 70 mm
150 x 95 mm
Quantity:
10 Pk.
6 Pk.
Unit Size:
Pack of 100
Pack of 120
Product Code. 17118660
Brand: 3M™ 7000097787
Only null left
Add to basket
Add to basket
Unique side grooves protect the fingernails, while also allowing for a better grip for more cleaning leverage. This pad protects fingernails for worry free washing up, reducing the need for gloves and the likelihood of broken nails.
  • Cleaning sponges with a unique ridge to protect finger nails
  • Handy size for a good grip and excellent durability
  • Foam shape is easy to handle
  • Reduces the need for gloves
  • Extremely long lasting
TRUSTED_SUSTAINABILITY
Specifications
Product Type Sponge
Color Yellow, White
Dimensions (L x W) 130 x 70 mm
For Use With (Application) Cleaning
Length (Metric) 130 mm
Material Foam
Quantity 10 Pk.
Style Nailsaver
Width (Metric) 70 mm