Learn More
Naphthalene, 99+%, scintillation grade, Thermo Scientific Chemicals
29.15 EUR - 179.90 EUR
Chemical Identifiers
CAS | 91-20-3 |
---|---|
Molecular Formula | C10H8 |
Molecular Weight (g/mol) | 128.17 |
MDL Number | MFCD00001742 |
InChI Key | UFWIBTONFRDIAS-UHFFFAOYSA-N |
PubChem CID | 931 |
ChEBI | CHEBI:16482 |
IUPAC Name | naphthalene |
SMILES | C1=CC2=CC=CC=C2C=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10417960
|
Thermo Scientific Acros
180200050 |
5 g | Glass Bottle |
29.15 EUR
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10792601
|
Thermo Scientific Acros
180202500 |
250 g | Glass Bottle |
65.12 EUR
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10124390
|
Thermo Scientific Acros
180200010 |
1 kg | Glass Bottle |
179.90 EUR
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For fluorescence/luminescence spectroscopyChemical Identifiers
91-20-3 | |
128.17 | |
UFWIBTONFRDIAS-UHFFFAOYSA-N | |
CHEBI:16482 | |
C1=CC2=CC=CC=C2C=C1 |
C10H8 | |
MFCD00001742 | |
931 | |
naphthalene |
Specifications
218.0°C | |
91-20-3 | |
99.0 | |
0.9900g/mL | |
78°C | |
Authentic | |
C10H8 | |
05, IV, 1640 | |
Solubility in water: 30 mg/l (25°C). Other solubilities: soluble in ethanol,, very soluble in acetone, chloroform, ether,, benzene, toluene, carbon disulfide | |
C1=CC2=CC=CC=C2C=C1 | |
128.17 | |
CHEBI:16482 | |
≥99% | |
Naphthalene, 99+% |
79.0°C to 82.0°C | |
Powder | |
100.0 | |
Beige to Red/Salmon | |
99+% | |
C10H8 | |
MFCD00001742 | |
15, 6455 | |
UFWIBTONFRDIAS-UHFFFAOYSA-N | |
naphthalene | |
931 | |
128.17 | |
0.99 |
Safety and Handling
GHS H Statement:
Suspected of causing cancer.
Very toxic to aquatic life with long lasting effects.
Harmful if swallowed.
GHS P Statement:
Use personal protective equipment as required.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep container tightly closed.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 202-049-5
missing translation for 'rtecsNumber' : QJ0525000
missing translation for 'tsca' : TSCA
- Naphthalene
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 2
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
- H302-Harmful if swallowed.
- H351-Suspected of causing cancer.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
Supplemental information
- MIXTURE LIST-Contains : Naphthalene
RUO – Research Use Only