missing translation for 'onlineSavingsMsg'
Learn More

Naphthalene, Scintillation Grade, ≥99%, MP Biomedicals™

Product Code. 11452462 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 g
Unit Size:
100g
This item is not returnable. View return policy

Product Code. 11452462

Brand: MP Biomedicals undefined

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 91-20-3
Color White
Assay Percent Range ≥99%
Molecular Formula C10H8
MDL Number MFCD00001742
Quantity 100 g
Synonym naphthalin, naphthene, camphor tar, tar camphor, white tar, albocarbon, naphthaline, moth flakes, moth balls, naphtalene
InChI Key UFWIBTONFRDIAS-UHFFFAOYSA-N
SMILES C1=CC2=CC=CC=C2C=C1
IUPAC Name naphthalene
Molecular Weight (g/mol) 128.17
PubChem CID 931
ChEBI CHEBI:16482
Formula Weight 128.2
Grade Scintillation
Physical Form Crystalline Powder
Chemical Name or Material Naphthalene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.