missing translation for 'onlineSavingsMsg'
Learn More

Naphthol As-D Chloroacetate, MP Biomedicals™

Product Code. p-7083104 Shop All MP Biomedicals Products
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 11463670

Brand: MP Biomedicals 02102424.1

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Naphthol As-D Chloroacetate, MP Biomedicals is a substrate for histochemical localization of esterases.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35245-26-2
Molecular Formula C20H16ClNO3
Molecular Weight (g/mol) 353.802
InChI Key FMVKYSCWHDVMGO-UHFFFAOYSA-N
Synonym naphthol as-d chloroacetate, 2-o-tolylcarbamoyl naphthalen-3-yl 2-chloroacetate, 3-hydroxy-2-naphthoic-o-toluidide chloroacetate, naphthol as-d-chloroacetate, 3-2-methylphenyl carbamoyl-2-naphthyl chloroacetate, 3-2-methylphenyl carbamoyl naphthalen-2-yl 2-chloroacetate, acetic acid, 2-chloro-,3-2-methylphenyl amino carbonyl-2-naphthalenyl ester, naphthol-as-d-chloroacetate, acetic acid, chloro-, s, naphthol as-d chloroacetate solution
PubChem CID 160247
IUPAC Name [3-[(2-methylphenyl)carbamoyl]naphthalen-2-yl] 2-chloroacetate
SMILES CC1=CC=CC=C1NC(=O)C2=CC3=CC=CC=C3C=C2OC(=O)CCl

Specifications

Melting Point 161°C to 171°C
Quantity 100 mg
Solubility Information Soluble in ethyl acetate (5mg/mL, clear and slightly yellow solution); methanol (1mg/mL, clear and colorless solution) and acetone.
Formula Weight 353.8
Physical Form Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.