Learn More
Whirl-Pak™ Write-On Scoop Sterile Sample Bag
Sterile scoop already contained inside the bag eliminates the need to carry two items to collect a sample and minimizes contamination.
Brand: Whirl-Pak™ B01350WA
Description
Sterile scoop is already contained inside the bag, eliminating the need to carry two items to collect the sample and minimizing contamination. The 2 oz. (60 cc) polypropylene scoop measures 7 in. (18.5 cm) long.
Capacity is measured in fluid ounces when the bag is closed and tab is folded over three times. Volume and dimensions are approximate, bags should not be used at temperatures above 180°F (82°C). Bags can be frozen to any temperature, even liquid nitrogen temperatures down to -346°F (-210°C) - careful handling required after freezing. All bags are sterilized after manufacturing.
Specifications
Sample Bag | |
Tape and Wire | |
Single-use | |
Sterile | |
Yes | |
23 x 11.5 cm (9 x 4.5 in.) | |
23 cm | |
11.5 cm |
Not Autoclavable | |
Clear with White Block | |
Polyethylene | |
0.064 mm | |
2.50 mil | |
9 in. | |
4.5 in. |