Learn More
Deutsch Neumann™ Rubber Policemen
Provides for quick and easy transfer of residues to glass surfaces. Deutsch Neumann™ Rubber Policemen are manufactured from natural rubber.
Brand: Deutsch Neumann™ 2250001
Additional Details : Weight : 0.00090kg
Description
- 5mm hole for insert
- Available in grey or red color
- Durometer Shore A 40
- Excellent tensile strength and resilience
Specifications
Rubber | |
Gray |